Learn More
Abnova™ Human ELA2B Full-length ORF (NP_056933.2, 1 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051032-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Like most of the human elastases, elastase 2B is secreted from the pancreas as a zymogen. In other species, elastase 2B has been shown to preferentially cleave proteins after leucine, methionine, and phenylalanine residues. [provided by RefSeq]
Sequence: MIRTLLLSTLVAGALSCGVSTYAPDMSRMLGGEEARPNSWPWQVSLQYSSNGQWYHTCGGSLIANSWVLTAAHCISSSGIYRVMLGQHNLYVAESGSLAVSVSKIVVHKDWNSDQVSKGNDIALLKLANPVSLTDKIQLACLPPAGTILPNNYPCYVTGWGRLQTNGALPDDLKQGRLLVVDYATCSSSGWWGSTVKTNMICAGGDGVICTCNGDSGGPLNCQASDGRWEVHGIGSLTSVLGCNYYYKPSIFTRVSNYNDWINSVIANNSpecifications
NP_056933.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ELA2B/MGC142100/MGC142102/MGC97052 | |
RP11-265F14.2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51032 | |
ELA2B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIRTLLLSTLVAGALSCGVSTYAPDMSRMLGGEEARPNSWPWQVSLQYSSNGQWYHTCGGSLIANSWVLTAAHCISSSGIYRVMLGQHNLYVAESGSLAVSVSKIVVHKDWNSDQVSKGNDIALLKLANPVSLTDKIQLACLPPAGTILPNNYPCYVTGWGRLQTNGALPDDLKQGRLLVVDYATCSSSGWWGSTVKTNMICAGGDGVICTCNGDSGGPLNCQASDGRWEVHGIGSLTSVLGCNYYYKPSIFTRVSNYNDWINSVIANN | |
RUO | |
RP11-265F14.2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |