missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ELA2B Full-length ORF (NP_056933.2, 1 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_056933.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51032 |
Molecular Weight (g/mol) | 55.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149542
|
Abnova™
H00051032-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139542
|
Abnova™
H00051032-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Like most of the human elastases, elastase 2B is secreted from the pancreas as a zymogen. In other species, elastase 2B has been shown to preferentially cleave proteins after leucine, methionine, and phenylalanine residues. [provided by RefSeq]
Sequence: MIRTLLLSTLVAGALSCGVSTYAPDMSRMLGGEEARPNSWPWQVSLQYSSNGQWYHTCGGSLIANSWVLTAAHCISSSGIYRVMLGQHNLYVAESGSLAVSVSKIVVHKDWNSDQVSKGNDIALLKLANPVSLTDKIQLACLPPAGTILPNNYPCYVTGWGRLQTNGALPDDLKQGRLLVVDYATCSSSGWWGSTVKTNMICAGGDGVICTCNGDSGGPLNCQASDGRWEVHGIGSLTSVLGCNYYYKPSIFTRVSNYNDWINSVIANNSpecifications
NP_056933.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MIRTLLLSTLVAGALSCGVSTYAPDMSRMLGGEEARPNSWPWQVSLQYSSNGQWYHTCGGSLIANSWVLTAAHCISSSGIYRVMLGQHNLYVAESGSLAVSVSKIVVHKDWNSDQVSKGNDIALLKLANPVSLTDKIQLACLPPAGTILPNNYPCYVTGWGRLQTNGALPDDLKQGRLLVVDYATCSSSGWWGSTVKTNMICAGGDGVICTCNGDSGGPLNCQASDGRWEVHGIGSLTSVLGCNYYYKPSIFTRVSNYNDWINSVIANN | |
RUO | |
RP11-265F14.2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51032 | |
ELA2B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ELA2B/MGC142100/MGC142102/MGC97052 | |
RP11-265F14.2 | |
Recombinant | |
wheat germ expression system |