Learn More
Human DC-SIGN+DC-SIGNR Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 8 publications
Brand:  R&D Systems MAB1621-100
 View more versions of this product
Product Code. 18758749
  • ONLINE EXCLUSIVE PRICE
    5050.00 NOK
    4773.30 NOK / 100µg
    Save 276.70 NOK
    5% Off
Description
DC-SIGN+DC-SIGNR Monoclonal specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DC-SIGN+DC-SIGNR
Monoclonal
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Q9H2X3
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
30835
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Flow Cytometry, Immunohistochemistry, CyTOF
120612
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD209
NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR Accession # Q9H2X3
100 μg
Primary
Recognizes both human DC-SIGN and human DC-SIGNR on transfected cells. Does not react with parental mouse cells or irrelevant transfectants.
Human
Supernatant