missing translation for 'onlineSavingsMsg'
Learn More

Human DC-SIGN+DC-SIGNR Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-5316914
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18792247 25 μg 25µg
18758749 100 μg 100µg
18710618 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18792247 Supplier R&D Systems Supplier No. MAB1621SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

DC-SIGN+DC-SIGNR Monoclonal specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC-SIGN+DC-SIGNR
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 120612
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H2X3
Gene Symbols CD209
Host Species Mouse
Immunogen NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR Accession # Q9H2X3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30835
Test Specificity Recognizes both human DC-SIGN and human DC-SIGNR on transfected cells. Does not react with parental mouse cells or irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.