missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DACT2 Full-length ORF (AAH92498.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00168002-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MWTPGGPPGSAGWDRRRLGARLRAAFAGLQELQGLRATQQERVRGALALQPPPAPAAPCGPHGLHGPEQQLEAALAALQEQLSRLRQQDIGLKTHLDQLDLQISKLQLDVGTASGEALDSDSRPSSGFYEMSDGGSCFLSTSCASVCSDHISPSLGSLLPVAQAHKARPSMGDWRPRSVDETTVPAWRPQATEEGARPPGSVEDAGQPWGTFWPRPVSTELCNAPGELDMHAPPAGCTSSSLTGVGSGLRGKCGLCGCQLPFCSVNTSSKTKSSGISRQQSpecifications
AAH92498.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf116/DAPPER2/DPR2/FLJ31232/MGC133141/MGC133142/RP11-503C24.7/bA503C24.7 | |
DACT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
168002 | |
DACT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWTPGGPPGSAGWDRRRLGARLRAAFAGLQELQGLRATQQERVRGALALQPPPAPAAPCGPHGLHGPEQQLEAALAALQEQLSRLRQQDIGLKTHLDQLDLQISKLQLDVGTASGEALDSDSRPSSGFYEMSDGGSCFLSTSCASVCSDHISPSLGSLLPVAQAHKARPSMGDWRPRSVDETTVPAWRPQATEEGARPPGSVEDAGQPWGTFWPRPVSTELCNAPGELDMHAPPAGCTSSSLTGVGSGLRGKCGLCGCQLPFCSVNTSSKTKSSGISRQQ | |
RUO | |
DACT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |