missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DACT2 Full-length ORF (AAH92498.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH92498.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 168002
Molecular Weight (g/mol) 55.8kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16199073
View Documents Promotion Details
Abnova™
H00168002-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16189073
View Documents Promotion Details
Abnova™
H00168002-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MWTPGGPPGSAGWDRRRLGARLRAAFAGLQELQGLRATQQERVRGALALQPPPAPAAPCGPHGLHGPEQQLEAALAALQEQLSRLRQQDIGLKTHLDQLDLQISKLQLDVGTASGEALDSDSRPSSGFYEMSDGGSCFLSTSCASVCSDHISPSLGSLLPVAQAHKARPSMGDWRPRSVDETTVPAWRPQATEEGARPPGSVEDAGQPWGTFWPRPVSTELCNAPGELDMHAPPAGCTSSSLTGVGSGLRGKCGLCGCQLPFCSVNTSSKTKSSGISRQQ
Specifications

Specifications

AAH92498.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.8kDa
Glutathione Sepharose 4 Fast Flow
MWTPGGPPGSAGWDRRRLGARLRAAFAGLQELQGLRATQQERVRGALALQPPPAPAAPCGPHGLHGPEQQLEAALAALQEQLSRLRQQDIGLKTHLDQLDLQISKLQLDVGTASGEALDSDSRPSSGFYEMSDGGSCFLSTSCASVCSDHISPSLGSLLPVAQAHKARPSMGDWRPRSVDETTVPAWRPQATEEGARPPGSVEDAGQPWGTFWPRPVSTELCNAPGELDMHAPPAGCTSSSLTGVGSGLRGKCGLCGCQLPFCSVNTSSKTKSSGISRQQ
RUO
DACT2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
168002
DACT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf116/DAPPER2/DPR2/FLJ31232/MGC133141/MGC133142/RP11-503C24.7/bA503C24.7
DACT2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications