missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Collectin-12/CL-P1 ELISA Kit

Product Code. 17104403
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17104403

Brand: Invitrogen™ EH121RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Collectin-12/CL-P1 quantitates human Collectin-12/CL-P1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Collectin-12/CL-P1.

This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Clp1; CL-P1; Colec12; collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; SCARA4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; SRCL
Assay Range 0.22 to 50 ng/mL
Assay Sensitivity 0.22 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 81035
Gene Symbol Colec12
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target COLEC12
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.