Learn More
Abnova™ Human CHRNB1 Partial ORF (NP_000738, 24 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001140-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome. [provided by RefSeq]
Sequence: SEAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALDSpecifications
NP_000738 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALD | |
RUO | |
CHRNB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1140 | |
CHRNB1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACHRB/CHRNB/CMS1D/CMS2A/SCCMS | |
CHRNB1 | |
Recombinant | |
wheat germ expression system |