missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHRNB1 Partial ORF (NP_000738, 24 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_000738 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1140 |
Molecular Weight (g/mol) | 37.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16186314
|
Abnova™
H00001140-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16196314
|
Abnova™
H00001140-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome. [provided by RefSeq]
Sequence: SEAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALDSpecifications
NP_000738 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACHRB/CHRNB/CMS1D/CMS2A/SCCMS | |
CHRNB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1140 | |
CHRNB1 (Human) Recombinant Protein (Q01) | |
SEAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALD | |
RUO | |
CHRNB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |