Learn More
R&D Systems™ Human Chitinase 3-like 1/YKL-40 Quantikine ELISA Kit
Product Code. 18732264
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18732264
Brand: R&D Systems™ DC3L10
Human Chitinase 3-like 1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Chitinase 3-like 1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Chitinase 3-like 1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 62.50 to 4000 pg/mL
Assay Sensitivity 8.15 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human CHI3L1
For Use With (Application) ELISA
Gene ID (Entrez) 1116
Gene Symbol CHI3L1
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40
Target Chitinase 3-like 1