missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD 74 ELISA Kit

Product Code. p-7219737
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Product Code. Quantity unitSize
17165309 5 x 96 Tests Each
17104833 96 Tests Pack of 96
2 options
This item is not returnable. View return policy

Product Code. 17165309

Brand: Invitrogen™ EH77RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human CD 74 quantitates human CD 74 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD 74.

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; DHLAG; dinucleotide microsatellite; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-GAMMA; Ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; Ii; invariant polypeptide of major histocompatibility complex, class II antigen-associated; INVG34; MHC class II-associated invariant chain; MHC HLA-DR gamma chain; p33
Assay Range 0.10 to 25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 972
Gene Symbol CD74
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD74
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.