Learn More
Invitrogen™ Human Cathepsin D ELISA Kit
Product Code. 17135309
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Product Code. 17135309
Brand: Invitrogen™ EH74RBX5
ELISA
Human Cathepsin D quantitates human Cathepsin D in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cathepsin D.
Cathepsin D has been reported to be involved in malignant tumor progression and with prognosis of various human cancers.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number CatD; Cathepsin D; Cathepsin D heavy chain; Cathepsin D light chain; CathepsinD; CD; ceroid-lipofuscinosis, neuronal 10; CLN10; CPSD; CTSD; epididymis secretory sperm binding protein Li 130 P; HEL-S-130 P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692
Assay Range 0.049 to 12 ng/mL
Assay Sensitivity 0.049 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1509
Gene Symbol Ctsd
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cathepsin D
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins