Learn More
Invitrogen™ Human Cathepsin D ELISA Kit
Product Code. 16627113
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Product Code. 16627113
Brand: Invitrogen™ EH74RB
ELISA
Human Cathepsin D quantitates human Cathepsin D in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cathepsin D.
Cathepsin D has been reported to be involved in malignant tumor progression and with prognosis of various human cancers.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number CatD; Cathepsin D; Cathepsin D heavy chain; Cathepsin D light chain; CathepsinD; CD; ceroid-lipofuscinosis, neuronal 10; CLN10; CPSD; CTSD; epididymis secretory sperm binding protein Li 130 P; HEL-S-130 P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692
Assay Range 0.049 to 12 ng/mL
Assay Sensitivity 0.049 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1509
Gene Symbol Ctsd
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cathepsin D
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins