missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human BTLA (NP_001078826) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Beschreibung
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human BTLA (NP_001078826) VersaClone cDNA clone contains the complete ORF for BTLA/CD272, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The BTLA/CD272 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature
Spezifikation
Spezifikation
| Gene | BTLA/CD272 |
| Accession Number | NP_001078826 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.87 |
| Insert Type | 739bp |
| Gene ID (Entrez) | 151888 |
| Gene Alias | B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only. Not for use in Humans.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur