missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ANO1/TMEM16A (NP_060513) VersaClone cDNA

Product Code. 18709618
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18709618 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18709618 Supplier R&D Systems™ Supplier No. RDC2500
  • ONLINE EXCLUSIVE PRICE
    3530.00 NOK
    3336.90 NOK / 10µg
    Save 193.10 NOK
    5% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human ANO1/TMEM16A (NP_060513) VersaClone cDNA clone contains the complete ORF for DOG1/TMEM16A, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The DOG1/TMEM16A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gene hANO1/TMEM16A
Accession Number NP_060513
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 55107
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.