missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACOX3 Full-length ORF (AAH17053.1, 1 a.a. - 624 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008310-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192352

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acyl-Coenzyme A oxidase 3 also know as pristanoyl -CoA oxidase (ACOX3)is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined. [provided by RefSeq]

Sequence: MASTVEGGDTALLPEFPRGPLDAYRARASFSWKELALFTEGEGMLRFKKTIFSALENDPLFARSPGADLSLEKYRELNFLRCKRIFEYDFLSVEDMFKSPLKVPALIQCLGMYDSSLAAKYLLHSLVFGSAVYSSGSERHLTYIQKIFRMEIFGCFALTELSHGSNTKAIRTTAHYDPATEEFIIHSPDFEAAKFWVGNMGKTATHAVVFAKLCVPGDQCHGLHPFIVQIRDPKTLLPMPGVMVGDIGKKLGQNGLDNGFAMFHKVRVPRQSLLNRMGDVTPEGTYVSPFKDVRQRFGASLGSLSSGRVSIVSLAILNLKLAVAIALRFSATRRQFGPTEEEEIPVLEYPMQQWRLLPYLAAVYALDHFSKSLFLDLVELQRGLASGDRSARQAELGREIHALASASKPLASWTTQQGIQECREACGGHGYLAMNRLGVLRDDNDPNCTYEGDNNILLQQTSNYLLGLLAHQVHDGACFRSPLKSVDFLDAYPGILDQKFEVSSVADCLDSAVALAAYKWLVCYLLRETYQKLNQEKRSGSSDFEARNKCQVSHGRPLALAFVELTVVQRFHEHVHQPSVPPSLRAVLGRLSALYALWSLSRHAALLYRAERRCSCPGRRDRSS
Specifications

Specifications

AAH17053.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ACOX3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8310
ACOX3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACOX3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers