Learn More
Abnova™ Human ACOX3 Full-length ORF (AAH17053.1, 1 a.a. - 624 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH17053.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8310 |
Molecular Weight (g/mol) | 96kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182352
|
Abnova™
H00008310-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16192352
|
Abnova™
H00008310-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Acyl-Coenzyme A oxidase 3 also know as pristanoyl -CoA oxidase (ACOX3)is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined. [provided by RefSeq]
Sequence: MASTVEGGDTALLPEFPRGPLDAYRARASFSWKELALFTEGEGMLRFKKTIFSALENDPLFARSPGADLSLEKYRELNFLRCKRIFEYDFLSVEDMFKSPLKVPALIQCLGMYDSSLAAKYLLHSLVFGSAVYSSGSERHLTYIQKIFRMEIFGCFALTELSHGSNTKAIRTTAHYDPATEEFIIHSPDFEAAKFWVGNMGKTATHAVVFAKLCVPGDQCHGLHPFIVQIRDPKTLLPMPGVMVGDIGKKLGQNGLDNGFAMFHKVRVPRQSLLNRMGDVTPEGTYVSPFKDVRQRFGASLGSLSSGRVSIVSLAILNLKLAVAIALRFSATRRQFGPTEEEEIPVLEYPMQQWRLLPYLAAVYALDHFSKSLFLDLVELQRGLASGDRSARQAELGREIHALASASKPLASWTTQQGIQECREACGGHGYLAMNRLGVLRDDNDPNCTYEGDNNILLQQTSNYLLGLLAHQVHDGACFRSPLKSVDFLDAYPGILDQKFEVSSVADCLDSAVALAAYKWLVCYLLRETYQKLNQEKRSGSSDFEARNKCQVSHGRPLALAFVELTVVQRFHEHVHQPSVPPSLRAVLGRLSALYALWSLSRHAALLYRAERRCSCPGRRDRSSSpecifications
AAH17053.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACOX3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8310 | |
ACOX3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RUO | |
ACOX3 | |
Recombinant | |
wheat germ expression system |