Learn More
HSPB2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21755-1-AP-150UL
 View more versions of this product
Product Code. 16861355
  • 4836.10 NOK / 150µL
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintece of muscle structure and function.
Specifications
Specifications
HSPB2
Polyclonal
Unconjugated
HSPB2
heat shock 27kDa protein 2, HSPB2
Rabbit
Antigen Affinity Chromatography
RUO
161476, 3316
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35878, Q16082
HSPB2
HSPB2 Fusion Protein Ag16146
150 μL
Primary
Human, Rat