missing translation for 'onlineSavingsMsg'
Learn More

HSPB2 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. p-7209673 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16871355 20 μL 20µL
16861355 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16871355

Brand: Proteintech 217551AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintece of muscle structure and function.

Specifications

Antigen HSPB2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSPB2
Gene Accession No. O35878, Q16082
Gene Alias heat shock 27kDa protein 2, HSPB2
Gene Symbols HSPB2
Host Species Rabbit
Immunogen HSPB2 Fusion Protein Ag16146
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161476, 3316
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.