Learn More
Proteintech HRP-conjugated Streptavidin
Shop All Proteintech Products
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
Description
The protein Streptavidin is purified from Streptomyces avidinii.
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 200 μL |
| Form | Liquid |