Learn More
HRG 293T Cell Overexpression Lysate (Denatured), Abnova
Product Code. 16146391
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 16146391
Brand: Abnova H00003273T01.100uL
Western Blotting
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description HRG 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_000412
Molecular Weight (g/mol) 57.86
pH Range 6.8
Plasmid pCMV-HRG full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-HRG antibody (H00003273-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL HRG 293T Cell Transient Overexpression Lysat
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol