Learn More
SCAT™ HPLC Waste Sets, V2.0
Product Code. 17875704 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
2.50 L
5 L
10 L
12 L
20 L
30 L
Material:
HDPE
HDPE-EC
Polypropylene
UN-X HDPE
Thread Size:
GL45
S50
S51
S55
S60/61
S70/71
S90
No. of Ports:
3 (2 Capillary, 1 Tubing)
5 (4 Capillary, 1 Tubing)
6 (3 Capillary, 3 Tubing)
Unit Size:
1 set
Each
| Product Code. | Capacity (Metric) | Material | Thread Size | No. of Ports | unitSize |
|---|---|---|---|---|---|
| 17875704 | 5 L | HDPE-EC | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 16493789 | 5 L | HDPE | GL45 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 16403799 | 10 L | HDPE | GL45 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17165501 | 5 L | Polypropylene | S50 | 3 (2 Capillary, 1 Tubing) | Each |
| 17385343 | 5 L | UN-X HDPE | S60/61 | 6 (3 Capillary, 3 Tubing) | 1 set |
| 17818733 | 12 L | HDPE | S60/61 | 3 (2 Capillary, 1 Tubing) | Each |
| 17835704 | 10 L | HDPE | S90 | 5 (4 Capillary, 1 Tubing) | 1 set |
| 17819273 | 2.50 L | HDPE | GL45 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17845714 | 5 L | HDPE | S55 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17825704 | 5 L | UN-X HDPE | S55 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17895714 | 5 L | UN-X HDPE | S51 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17885714 | 20 L | HDPE | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17875714 | 5 L | UN-X HDPE | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17855714 | 20 L | HDPE | S70/71 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17865714 | 5 L | HDPE-EC | S50 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17825724 | 10 L | HDPE-EC | S50 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17845704 | 10 L | HDPE-EC | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17855704 | 20 L | HDPE-EC | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17805724 | 30 L | HDPE-EC | S60/61 | 3 (2 Capillary, 1 Tubing) | 1 set |
| 17815724 | 10 L | HDPE-EC | S90 | 5 (4 Capillary, 1 Tubing) | 1 set |
| 18606478 | 5 L | UN-X HDPE | S60/61 | 6 (3 Capillary, 3 Tubing) | 1 set |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
- Customized system with high-quality set components
- Safety canisters made of HDPE or polypropylene material
- Attractive price compared to single purchase
- Ready to use, no further components necessary
Specifications
Specifications
| For Use With (Application) | HPLC Systems |
| Type | Complete Solvent Collection System |
| Solvent Channels | 3 |
| Material | HDPE-EC |
| Includes | Safety Waste Cap, PTFE-EL, 2 x PFA fitting 2.3 mm OD, 2 x PFA fitting 3.2 mm OD, 1 x PP tube connector 5 to 11.5 mm ID, 1 x GL14 connection for exhaust filter, 1 x canister, S60/61, 5 L, PE-HD elektrostatic conductive, 1 x grounding cable, 2 x clamp, length 1.5 m, 1 x exhaust filter S, with splash guard and change indicator |
| Color | Black |
| Thread Size | S60/61 |
| Capacity (Metric) | 5 L |
| No. of Ports | 3 (2 Capillary, 1 Tubing) |