Learn More
SCAT™ HPLC Waste Sets, V2.0
Product Code. 17165501 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
2.50 L
5 L
10 L
12 L
20 L
30 L
Material:
HDPE
HDPE-EC
Polypropylene
UN-X HDPE
Thread Size:
GL45
S50
S51
S55
S60/61
S70/71
S90
No. of Ports:
3 (2 Capillary, 1 Tubing)
5 (4 Capillary, 1 Tubing)
6 (3 Capillary, 3 Tubing)
Unit Size:
1 set
Each
Product Code. 17165501
Brand: SCAT™ 307328
Only null left
Add to basket
Add to basket
HPLC Waste Sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT™ SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.
SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
  • Customized system with high-quality set components
  • Safety canisters made of HDPE or polypropylene material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary
Specifications
For Use With (Equipment) HPLC Systems
Type Complete Solvent Collection System
Solvent Channels 3
Material Polypropylene
Includes Safety Waste Cap, 2 x 3.2 mm OD, 1 x 5 to 11.5 mm ID, Polypropylene Canister, Exhaust Filter S with Splash Guard and Exchange Indicator
Color White
Thread Size S50
Capacity (Metric) 5 L
No. of Ports 3 (2 Capillary, 1 Tubing)