missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMEZ Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23965-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May function as a transcriptional regulator.Specifications
HOMEZ | |
Polyclonal | |
Unconjugated | |
HOMEZ | |
HOMEZ, KIAA1443 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
260325, 57594 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IX15, Q8K3E9 | |
Homez | |
HOMEZ Fusion Protein Ag21090 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |