missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMEZ Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | HOMEZ |
---|---|
Concentration | 0.55 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802565
|
Proteintech
23965-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16892555
|
Proteintech
23965-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
May function as a transcriptional regulator.Specifications
HOMEZ | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human | |
Q8IX15, Q8K3E9 | |
260325, 57594 | |
HOMEZ Fusion Protein Ag21090 | |
Primary | |
-20°C | |
HOMEZ |
0.55 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HOMEZ, KIAA1443 | |
Homez | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |