Learn More
WTW™ Holding Clip for WTW™ SenTix™ pH Electrodes and Temperature Probe
Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor
Brand: WTW™ 109821
Additional Details : Weight : 0.01000kg
Description
pH and ORP measurements must always have a temperature indication. During pH measurement, the indication of a temperature to compensate the temperature effect, and the documentation, with ORP measurement, only for documentation purposes. During ISE measurement it is important that the calibration and the measurement are performed in the same temperature range. Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor. The temperature sensors are equipped with two 4 mm banana plugs.Specifications
Holding Clip | |
pH combination electrode and temperature sensor TFK 325 |