missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | HMGB3 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885635
|
Proteintech
27465-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875635
|
Proteintech
27465-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2 , HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.Specifications
HMGB3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O15347 | |
3149 | |
HMGB3 Fusion Protein Ag26361 | |
Primary | |
-20°C | |
HMGB3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
high mobility group box 3, High mobility group protein 2a, High mobility group protein 4, High mobility group protein B3, HMG 2a, HMG 4, HMG2A, HMG4, HMGB3 | |
HMGB3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |