missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27465-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2 , HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.Specifications
HMGB3 | |
Polyclonal | |
Unconjugated | |
HMGB3 | |
high mobility group box 3, High mobility group protein 2a, High mobility group protein 4, High mobility group protein B3, HMG 2a, HMG 4, HMG2A, HMG4, HMGB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3149 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15347 | |
HMGB3 | |
HMGB3 Fusion Protein Ag26361 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |