missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA B7 Antibody (BB7.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
1995.00 NOK - 4730.00 NOK
Specifications
| Antigen | HLA B7 |
|---|---|
| Clone | BB7.1 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004916
|
Novus Biologicals
NB100-64159-0.025MG |
0.025 mg |
1995.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18059033
|
Novus Biologicals
NB100-64159 |
0.1 mg |
4730.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA B7 Monoclonal specifically detects HLA B7 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HLA B7 | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human, Primate | |
| ankylosing spondylitis, AS, b'DT, Bw-22, Bw-4, Bw-41, Bw-44, Bw-45, Bw-46, Bw-47, Bw-48, Bw-50, Bw-52, Bw-53, Bw-54, Bw-55, Bw-56, Bw-57, Bw-58, Bw-60, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-12 alpha chain, HLA class I histocompatibility antigen, B-21 alpha chain, HLA class I histocompatibility antigen, B-5 alpha chain, HLA class I histocompatibility antigen, B-7 alpha chain, HLAB, HLA-B27, HLA-B73, leukocyte antigen class I-B, lymphocyte antigen, major histocompatibility complex, class I, B, MGC111087, MHC class I antigen B*13, MHC class I antigen B*14, MHC class I antigen B*15, MHC class I antigen B*18, MHC class I antigen B*27, MHC class I antigen B*35, MHC class I antigen B*37, MHC class I antigen B*38, MHC class I antigen B*39, MHC class I antigen B*40, MHC class I antigen B*41, MHC class I antigen B*42, MHC class I antigen B*44, MHC class I antigen B*45, MHC class I antigen B*46, MHC class I antigen B*47, MHC class I anti | |
| HLA-B | |
| IgG1 | |
| Protein A purified | |
| Recognizes the HLA-B7 antigen and does not cross-react with HLA-B27 or other related antigens. It can be used to distinguish true HLA-B27 positives from false HLA-B27 positives (i.e. HLA-B7 positive) in the investigation of diseases such as ankylosing spondylitis and anterior uveitis. |
| BB7.1 | |
| Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P01889 | |
| 3106 | |
| Papain solubilized HLA-B7 antigen | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title