missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA B7 Antibody (BB7.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64159-0.025MG
This item is not returnable.
View return policy
Description
HLA B7 Monoclonal specifically detects HLA B7 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HLA B7 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200 | |
| P01889 | |
| HLA-B | |
| Papain solubilized HLA-B7 antigen | |
| 0.025 mg | |
| Primary | |
| Recognizes the HLA-B7 antigen and does not cross-react with HLA-B27 or other related antigens. It can be used to distinguish true HLA-B27 positives from false HLA-B27 positives (i.e. HLA-B7 positive) in the investigation of diseases such as ankylosing spondylitis and anterior uveitis. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| BB7.1 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| ankylosing spondylitis, AS, b'DT, Bw-22, Bw-4, Bw-41, Bw-44, Bw-45, Bw-46, Bw-47, Bw-48, Bw-50, Bw-52, Bw-53, Bw-54, Bw-55, Bw-56, Bw-57, Bw-58, Bw-60, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-12 alpha chain, HLA class I histocompatibility antigen, B-21 alpha chain, HLA class I histocompatibility antigen, B-5 alpha chain, HLA class I histocompatibility antigen, B-7 alpha chain, HLAB, HLA-B27, HLA-B73, leukocyte antigen class I-B, lymphocyte antigen, major histocompatibility complex, class I, B, MGC111087, MHC class I antigen B*13, MHC class I antigen B*14, MHC class I antigen B*15, MHC class I antigen B*18, MHC class I antigen B*27, MHC class I antigen B*35, MHC class I antigen B*37, MHC class I antigen B*38, MHC class I antigen B*39, MHC class I antigen B*40, MHC class I antigen B*41, MHC class I antigen B*42, MHC class I antigen B*44, MHC class I antigen B*45, MHC class I antigen B*46, MHC class I antigen B*47, MHC class I anti | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3106 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction