Learn More
Cytiva HiTrap™ Q Sepharose Fast Flow IEX Columns
Product Code. 11535025 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
25mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11535025 5 mL 25mL
10607275 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11535025 Supplier Cytiva Supplier No. 17515601
 Restricted Item
In Stock
Use for small-scale protein purifications. Cytiva Life Sciences™ HiTrap™ Q Sepharose Fast Flow IEX Columns are strong anion exchangers for rapid separations either alone or connected in series.
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential
TRUSTED_SUSTAINABILITY
Specifications
Description HiTrap Q FF
Diameter (English) Inner Column 0.6 in.
Flow Rate <20 mL/min.
For Use With (Application) HiTrap Q FF is a strong anion exchange chromatography column for small-scale protein purification.
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 5 mL
compliance-icons
Product Identifier
  • HiTrap Q FF 5 ml, 5 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol