Learn More
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns
Product Code. 10734572 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate unitSize
10734572 5 x 1 mL <4 mL/min. Pack of 5
11673074 5 x 5 mL <20 mL/min. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10734572 Supplier Cytiva Supplier No. 17547051
  • ONLINE EXCLUSIVE PRICE
    1860.00 NOK
    1776.00 NOK / Pack of 5
    Save 84.00 NOK
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
High-throughput medium for intermediate and polishing protein purification
  • Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
  • Screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Strong quartenary amine anion exchanger
TRUSTED_SUSTAINABILITY
Specifications
Description HiTrap Capto Q ImpRes, 5 x 1 mL
Flow Rate <4 mL/min.
For Use With (Application) HiTrap Capto Q ImpRes chromatography column is packed with a strong anion exchange modern resin for high resolution, small-scale protein purification.
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 7 mm
Volume (Metric) 1 mL
Quantity 5 x 1 mL
compliance-icons
Product Identifier
  • HiTrap Capto Q ImpRes 1 ml, 5 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.