Learn More
Cytiva HiTrap Capto MMC Columns
Product Code. 10480315 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Flow Rate:
<4 mL/min.
300kPa at 600cm/hr.
Max. Pressure:
3 bar
44 bar
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Diameter (English) Inner Flow Rate Max. Pressure Unit Size
10480315 5 x 1mL 0.27 in. <4 mL/min. 44 bar Pack of 5
11748408 5 x 5mL 0.62 in. 300kPa at 600cm/hr. 3 bar Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10480315 Supplier Cytiva Supplier No. 11003273
 Restricted Item
In Stock
HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.
  • High dynamic binding capacity at high conductivity.
  • Selectivity allows binding in the presence of salt.
  • High volume throughput.
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Aqueous Buffer
Product Type HiTrap Capto MMC column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Max. Pressure 44 bar
pH Range 2 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Bind Proteins at the conductivity of the feed material and to solve specific purification challenges
Show More Show Less
compliance-icons
Product Identifier
  • hitrap capto MMC
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.