Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic system
- High flowrates and good scale-up potential
Specifications
Specifications
| Description | HiTrap ANX FF (High Sub) |
| Diameter (English) Inner Column | 0.6 in. |
| Flow Rate | <20 mL/min. |
| For Use With (Application) | HiTrap ANX (high sub) FF is prepacked with ANX (high sub) Sepharose Fast Flow, and is a weak anion exchanger for small scale purification of higher molecular weight protein as well as screening of binding and elutions conditions. |
| Content And Storage | 4°C to 30°C in 20% Ethanol |
| Diameter (Metric) Inner Column | 16 mm |
| Volume (Metric) | 5 mL |
| Quantity | 5 x 5 mL |
Safety and Handling
Signal Word
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.