missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
2075.00 NOK - 4690.00 NOK
Specifications
| Antigen | Histone H3 |
|---|---|
| Concentration | 18.0mg/mL |
| Dilution | Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, ChIP Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18234200
|
Novus Biologicals
NB500-267SS |
0.025 mL |
2075.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769413
|
Novus Biologicals
NB500-267 |
4690.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Histone H3 | |
| Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Precipitation | |
| RUO | |
| Human, Mouse | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| H3C14 | |
| IgG | |
| Ammonium sulfate precipitation | |
| Human and mouse. Predicted to cross-react with all mammals and a wide range of other species due to sequence homology. |
| 18.0mg/mL | |
| Western Blot, ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| PBS with 0.02% Sodium Azide | |
| 126961 | |
| This Histone H3 antibody was raised against a synthetic peptide corresponding to a sequence within amino acids 100-135 of human Histone H3. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title