missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB500-267SS
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Histone H3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| H3C14 | |
| This Histone H3 antibody was raised against a synthetic peptide corresponding to a sequence within amino acids 100-135 of human Histone H3. | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human and mouse. Predicted to cross-react with all mammals and a wide range of other species due to sequence homology. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ChIP Assay | |
| 18.0mg/mL | |
| Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 35031618), Chromatin Immunoprecipitation (ChIP) | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Rabbit | |
| 15 kDa | |
| 0.025 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Human, Mouse | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction