missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2480.00 NOK - 4095.00 NOK
Specifications
| Antigen | Histone H2AX |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18716613
|
Novus Biologicals
NBP2-27427SS |
0.025 mg |
2480.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18104045
|
Novus Biologicals
NBP2-27427 |
0.1 mg |
4095.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Histone H2AX | |
| Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| P16104 | |
| 3014 | |
| A portion of amino acids 100-143 of human Histone H2A.x was used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| H2A.X, H2A/X, H2AFX | |
| H2AFX | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the Histone H2A.x antibody is 100% homologous in human and monkey, 94% homologous in cow and dog, 88% homologous in rat and 82% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title