missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27427SS
This item is not returnable.
View return policy
Description
Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Histone H2AX | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| H2A.X, H2A/X, H2AFX | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the Histone H2A.x antibody is 100% homologous in human and monkey, 94% homologous in cow and dog, 88% homologous in rat and 82% homologous in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml | |
| P16104 | |
| H2AFX | |
| A portion of amino acids 100-143 of human Histone H2A.x was used as the immunogen for the antibody. | |
| 0.025 mg | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific | |
| 3014 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction