compliance-icons
Cytiva HiPrep™ Heparin Fast Flow 16/10 Column
Product Code. 10657455 Shop All Cytiva Products
1 x 20 mL, Each
Click to view available options
Quantity:
1 x 20 mL
Unit Size:
Each
For preparative purification of proteins with affinity for heparin
  • Immobilized heparin is used for purification of antithrombin III, coagulation factors, and other plasma proteins
  • Compatible with simple pump-based configurations and other chromatography systems such as AKTA design
  • Optimized for reliable, reproducible separations
  • High loading capacity and fast purifications
  • Rapid initial capture of proteins from start material
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner Column 0.6 in.
Flow Rate 2 to 10 mL/min.
Content And Storage 4°C to 30°C, 0.05M Sodium Acetate containing 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 20 mL
Quantity 1 x 20 mL
compliance-icons
Product Identifier
  • HiPrep Heparin FF 16/10
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Product Title
Cytiva HiPrep™ Heparin Fast Flow 16/10 Column >