Learn More
Fisherbrand™ High Speed Module
Product Code. 17197804 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Blue Line 5 / IQ5
Blue Line 6 / IQ6
Unit Size:
Each
Product Code. 17197804
Only null left
Add to basket
Add to basket
High speed module consist of 2 tanks on top of the machine.
  • Tanks electrically heated
  • Water is preheated before going into the washing chamber, allowing for shorter cycle times
Specifications
Product Type High Speed Module
For Use With (Equipment) Blue Line 5 / IQ5
Includes 2 Tanks