Learn More
- Home
- Products
- Thermometers and Temperature Measurement
- Manual Thermometers
- Brannan™ Precision Lab Thermometer with 3-points Traceable Calibration
Brannan™ Precision Lab Thermometer with 3-points Traceable Calibration
Product Code. 17853308
Change view
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 201°C
-1°C to 250°C
-1°C to 51°C
-10°C to +110°C
-10°C to +150°C
-10°C to +50°C
-100°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
-50°C to +50°C
0°C to 40°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
20.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
27 product options available for selection
| Product Code. | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Unit Size |
|---|---|---|---|---|---|
| 17853308 | -100°C to +50°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17863298 | -20°C to +110°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17883018 | -1°C to 101°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17873298 | -1°C to 250°C | 1°C | Partial Immersion | 40.5 cm | Each |
| 17883038 | -50°C to +50°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17803308 | 0°C to 40°C | 0.1°C | Partial Immersion | 40.5 cm | Each |
| 17873038 | -10°C to +150°C | 1°C | Partial Immersion | 20.5 cm | Each |
| 17822988 | -1°C to 51°C | 0.1°C | Total Immersion | 40.5 cm | Each |
| 17813038 | -1°C to 51°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17803038 | -35°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17823018 | -1°C to 250°C | 1°C | Total Immersion | 40.5 cm | Each |
| 17873018 | 0°C to 40°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17893028 | 0°C to 40°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17823038 | -20°C to +110°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17833308 | -20°C to +150°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17843308 | -1°C to 201°C | 0.5°C | Partial Immersion | 40.5 cm | Each |
| 17853078 | -1°C to 51°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17883068 | -1°C to 101°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17843118 | -10°C to +150°C | 1°C | Total Immersion | 20.5 cm | Each |
| 17863048 | 0°C to 40°C | 0.1°C | Total Immersion | 40.5 cm | Each |
| 17813108 | -10°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17893298 | -35°C to +50°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17823308 | -20°C to +150°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17833118 | -10°C to +110°C | 1°C | Partial Immersion | 20.5 cm | Each |
| 17813308 | -1°C to 201°C | 0.5°C | Total Immersion | 40.5 cm | Each |
| 17823118 | -1°C to 51°C | 0.1°C | Partial Immersion | 40.5 cm | Each |
| 17853118 | -10°C to +110°C | 1°C | Total Immersion | 20.5 cm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- A range of spirit filled, high precision thermometers which are supplied with a calibration certificate
- Safe non-mercury filling
- Presented in a study cardboard cylinder for storage
- Made in UK
Specifications
Specifications
| Calibration Points | 0°C, 35°C |
| Certifications/Compliance | UK Traceable to National Standards ISO 9001 |
| Immersion Type | Total Immersion |
| Length (Metric) | 30.5 cm |
| Material | Glass |
| Thermometer Fill | Red Spirit |
| Max. Temperature (English) | 122°F |
| Max. Temperature (Metric) | 50°C |
| Min. Temperature (English) | -148°F |
| Min. Temperature (Metric) | -100°C |
| Temperature Range (English) | -148°F to +122°F |
| Temperature Range (Metric) | -100°C to +50°C |
| Accuracy | ± 1.0°C |
| Subdivision | 1°C |
| Width (Metric) | 1.2 cm |
| Show More Show Less |