missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ HI 3220-02 Model pH/mV/Temperature Bench Meter

Product Code. 10703407
Change view
Click to view available options
Temperature Range (Metric):
-20.0°C to +120.0°C
Unit Size:
Each
Product Code. Temperature Range (Metric) unitSize
10703407 -20.0°C to +120.0°C Each
1 options
This item is not returnable. View return policy

Product Code. 10703407

Brand: Hanna Instruments™ HI322002

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Professional bench meter designed to provide high accuracy and ease of use. Hanna Instruments™ HI 3220-02 Model pH/mV/Temperature Bench Meter can be used in the laboratory or in harsh industrial conditions.

  • One input channel
  • pH Calibration check
  • Five-point pH calibration with seven standard and five custom buffers
  • Stability, interval and log on demand logging
  • Up to 400 log on demand records and 600 automatic logging records
  • Messages on the graphic LCD for an easy and accurate calibration
  • Multi-language support
  • GLP features
  • Optional PC interface via USB

Specifications

Includes HI 1131B pH with BNC connector and HI 7662-T Stainless Steel temperature Probe, starter pack of buffer and cleaning solutions, 12Vdc adapter, Instructions
Accuracy (Temperature) ±0.2°C
Accuracy (pH) ±0.01, ±0.002
Range (pH) -2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999
Resolution (pH) 0.001, 0.01, 0.1
Range (ORP) ±2000mV
Calibration Points 1.68, 4.01, 6.86, 7.01, 9.18, 10.01, 12.45
Temperature Range (Metric) -20.0°C to +120.0°C
Resolution (Temperature) 0.1°C
Display Type LCD
Inputs 1
Weight (Metric) 1.8 kg
Model HI 3220-02
Power Supply 12VDC
Probe Type HI7662-T Stainless Steel temperature Probe with 1m cable
Temperature Compensation -20.0°C to +120.0°C
Channels 1
Type pH/ORP Benchtop Meter
Voltage 240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.