missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HFE2 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
1786.00 NOK - 4515.00 NOK
Specifications
| Antigen | HFE2 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17684226
|
Invitrogen™
MA543774 |
20 μL |
1786.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17674226
![]() |
Invitrogen™
MA543773 |
100 μL |
4515.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HFE2 is involved in iron metabolism. It may be a component of the signaling pathway which activates hepcidin or it may act as a modulator of hepcidin expression. It could also represent the cellular receptor for hepcidin. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. Defects in this gene are the cause of hemochromatosis type 2A, also called juvenile hemochromatosis (JH). JH is an early-onset autosomal recessive disorder due to severe iron overload resulting in hypogonadotrophic hypogonadism, hepatic fibrosis or cirrhosis and cardiomyopathy, occurring typically before age of 30.Specifications
| HFE2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 2310035L15Rik; 5230400G09Rik; AI414844; AI789733; DL M; DL-M; Fidgetin; haemojuvelin; HEMOCHROMATOSIS; hemochromatosis type 2 (juvenile); hemochromatosis type 2 (juvenile) (human homolog); hemochromatosis type 2 protein; Hemochromatosis type 2 protein homolog; hemojuvelin; Hemojuvelin BMP coreceptor; HFE 2; HFE2; HFE2A; HJV; JH; MGC23953; OTTHUMP00000059680; repulsive guidance molecule c; RGM C; RGM domain family member C; RGMC | |
| HFE2 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Q6ZVN8, Q8N7M5 | |
| 148738, 310681 | |
| Lys234-Ser416 (Accession Q6ZVN8), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HFE2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title