Learn More
Thermo Scientific™ HEPES Lysis Buffer with Triton™ X-100
Product Code. 15406019
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
Product Code. Quantity unitSize
15406019 500mL 500mL
15496009 250mL 250mL
2 options
Product Code. 15406019
Brand: Thermo Scientific™ J63860.AP
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500mL
Format Liquid
Product Type Cell Lysis Reagent
Content And Storage Ambient temperatures