Learn More
Fisherbrand™ HDPE Coated Wire Rack
Product Code. 16645834 Shop All Fisherbrand Products
Change view
Click to view available options
Array:
3 x 8
4 x 10
4 x 12
6 x 12
6 x 8
8 x 10
9 x 12
Size:
200 x 83 x 59 mm
204 x 161 x 65 mm
205 x 105 x 65 mm
237 x 127 x 65 mm
240 x 190 x 80 mm
240 x 90 x 80 mm
241 x 188 x 65 mm
273 x 105 x 80 mm
277 x 209 x 98 mm
280.4 x 120.8 x 81.6 mm
290 x 118 x 80 mm
290 x 230 x 80 mm
350 x 260 x 98 mm
357 x 139 x 98 mm
Diameter (Metric) Well:
10-13 mm
13-16 mm
16-20 mm
20-24 mm
20-25 mm
25-30 mm
30-40 mm
Unit Size:
Each
Product Code. Array Size Diameter (Metric) Well unitSize
16645834 8 x 10 240 x 190 x 80 mm 16-20 mm Each
16685834 3 x 8 273 x 105 x 80 mm 20-25 mm Each
16605844 3 x 8 357 x 139 x 98 mm 30-40 mm Each
16605834 4 x 10 200 x 83 x 59 mm 13-16 mm Each
16635834 4 x 10 240 x 90 x 80 mm 16-20 mm Each
16655834 4 x 10 280.4 x 120.8 x 81.6 mm 20-24 mm Each
16665834 4 x 10 290 x 118 x 80 mm 20-25 mm Each
16615834 4 x 12 237 x 127 x 65 mm 13-16 mm Each
16695834 6 x 8 277 x 209 x 98 mm 25-30 mm Each
16615844 6 x 8 350 x 260 x 98 mm 30-40 mm Each
16685824 6 x 12 205 x 105 x 65 mm 10-13 mm Each
16675834 8 x 10 290 x 230 x 80 mm 20-25 mm Each
16695824 9 x 12 204 x 161 x 65 mm 10-13 mm Each
16625834 9 x 12 241 x 188 x 65 mm 13-16 mm Each
14 options
Product Code. 16645834
Only null left
Add to basket
Add to basket
HDPE Coated Wire Rack is durable for transport or sample handling. It is chemically-resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
  • Chemically-resistant HDPE Coated Wire Racks
  • Racks can be used between temperatures of -40°C and 121°C
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
  • Autoclavable
  • Slotted Bottom Support
Specifications
Autoclavable Autoclavable
Color Blue
Holds Tubes
Material HDPE-coated Wire
Positions 1
Shape Rectangular, Rounded Corners
Stackable Yes, When Empty
Sterility Non-sterile
No. per Case 24
No. per Pack 1
Size 240 x 190 x 80 mm
Dimensions (L x W) 240 x 190 mm
Dimensions (L x W x H) 240 x 190 x 80 mm
Length (Metric) 240 mm
Length (English) 9.50 in.
Width (English) 7.60 in.
Height (English) 3.10 in.
Diameter (Metric) Well 16-20 mm
Product Type Tube Rack
Shelves 3 Tiers
Height (Metric) 80 mm
Array 8 x 10
Width (Metric) 190 mm