Learn More
SymLine Safety Waste Caps
GREENER_CHOICE
Product Code. 17192566
Change view
Click to view available options
Size:
GL 45, GL 25
GL45
S60/61
Includes:
2 x PFA-fittings
4 x PFA-fittings
4 x PFA-fittings, 1 x tube connector
7 x PFA-fittings
Electronic fill level control
Filling Lance, Connector for exhaust filter
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes unitSize
17192566 GL45 4 x PFA-fittings Each
16305988 GL 45, GL 25 Filling Lance, Connector for exhaust filter Each
17172566 GL45 2 x PFA-fittings Each
17102576 GL45 4 x PFA-fittings, 1 x tube connector Each
17182566 GL45 7 x PFA-fittings Each
10820411 S60/61 Electronic fill level control Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17192566 Supplier SymLine Supplier No. 306493
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
  • Safety Waste Caps are available with different thread sizes for attachment to the pipe system and with different numbers of hose connections for disposal of the liquid waste.
  • Optionally, Safety Waste Caps are available with electronic level sensors that indicate the degree of filling of the disposal container.
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
  • Specifications
    For Use With (Equipment) SymLine System for capillaries with (OD) 1.6/2.3/3.2 mm
    Includes 4 x PFA-fittings
    Material PTFE
    Size GL45
    For Use With (Application) Waste Disposal System
    Type Safety Waste Cap