Learn More
SymLine Safety Waste Caps
Description
Specifications
Specifications
| For Use With (Equipment) | SymLine System for connection table assembly |
| Includes | Electronic fill level control |
| Material | HDPE-EC |
| Size | S60/61 |
| For Use With (Application) | Waste Disposal System |
| Type | Safety Waste Cap |
| Color | Black |