missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAMP Monoclonal Antibody (D7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA543786
Additional Details : Weight : 0.01000kg
Description
HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.Specifications
HAMP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P81172 | |
HAMP | |
Ser25-Thr84 (Accession P81172), with N-terminal His Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
D7 | |
Unconjugated | |
HAMP | |
HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003 | |
Mouse | |
Protein A/G | |
RUO | |
57817 | |
-20°C | |
Liquid |