missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAMP Monoclonal Antibody (D7), Invitrogen™

Mouse Monoclonal Antibody
1834.00 NOK - 4635.00 NOK
Specifications
| Antigen | HAMP |
|---|---|
| Clone | D7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17614246
|
Invitrogen™
MA543787 |
20 μL |
1834.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17604246
![]() |
Invitrogen™
MA543786 |
100 μL |
4635.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.Specifications
| HAMP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003 | |
| HAMP | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| D7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P81172 | |
| 57817 | |
| Ser25-Thr84 (Accession P81172), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HAMP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title