Learn More
H-B Instrument™ Durac™ Refrigerator Thermometers
Product Code. 17541305
Change view
Click to view available options
Temperature Range (Metric):
-30°C to +30°C
Temperature Range (English):
-20°F to +80°F
Accuracy:
1°C, 2°F
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Temperature Range (English) Accuracy Material unitSize
17541305 -30°C to +30°C -20°F to +80°F 1°C, 2°F Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17541305 Supplier H-B Instrument™ Supplier No. B613202100
Add to basket
Add to basket
For measuring refrigerator/freezer temperatures
  • Available in stainless-steel or plastic
Specifications
Display Dimensions 76 mm
For Use With (Application) Measuring refrigerator/freezer temperatures
Material Stainless Steel
Max. Temperature (English) 80°F
Max. Temperature (Metric) 30°C
Min. Temperature (English) -20°F
Min. Temperature (Metric) -30°C
Temperature Range (English) -20°F to +80°F
Temperature Range (Metric) -30°C to +30°C
Accuracy 1°C, 2°F